missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RRAS Protein

A cDNA sequence encoding the RRAS was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP13363-10ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15983329

  • 203.00€ / 10µg
Expédition estimée: 17-01-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

RRAS Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MSSGAASGTG RGRPRGGGPG PGDPPPSETH KLVVVGGGGV GKSALTIQFI QSYFVSDYDP TIEDSYTKIC SVDGIPARLD ILDTAGQEEF GAMREQYMRA GHGFLLVFAI NDRQSFNEVG KLFTQILRVK DRDDFPVVLV GNKADLESQR QVPRSEASAF GASHHVAYFE ASAKLRLNVD EAFEQLVRAV RKYQEQELPP SPPSAPRKKG GGCPC
Greater than 85% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
RRAS Human solution (0.5 mg/1ml) containing 20mM Tris-HCl pH 8.0, 0.1M NaCl and 10% glycerol.
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions