missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human S100B Protein

A cDNA sequence encoding the S100B was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP13404-20ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15983459

  • 203.00€ / 20µg
Expédition estimée: 17-01-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

6285
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MSELEKAMVA LIDVFHQYSG REGDKHKLKK SELKELINNE LSHFLEEIKE QEVVDKVMET LDNDGDGECD FQEFMAFVAM VTTACHEFFE HE
Greater than 90.0% as determined by SDS-PAGE.
S100B Protein
Research Use Only
S100B
Human
His
S100B (1 mg/ml) is supplied in 20mM Tris-HCl buffer (pH 8.0), 1mM DTT and 10% glycerol.
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions