missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Semaphorin-3F Protein
A DNA sequence encoding the Homo sapiens (Human) Semaphorin-3F, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP10123-ma-50ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_001305729.1 | |
Semaphorin-3F Protein | |
Research Use Only | |
SEMA3F | |
Recombinant Protein | |
Mammal | |
SFPTQDHLPA TPRVRLSFKE LKATGTAHFF NFLLNTTDYR ILLKDEDHDR MYVGSKDYVL SLDLHDINRE PLIIHWAASP QRIEECVLSG KDVNGECGNF VRLIQPWNRT HLYVCGTGAY NPMCTYVNRG RRAQATPWTQ TQAVRGRGSR ATDGALRPMP TAPRQDYIFY LEPERLESGK GKCPYDPKLD TASALINEEL YAGVYIDFMG TDAAIFRTLG KQTAMRTDQY NSRWLNDPSF IHAELIPDSA ERNDDKLYFF FRERSAEAPQ SPAVYARIGR ICLNDDGGHC CLVNKWSTFL KARLVCSVPG EDGIETHFDE LQDVFVQQTQ DVRNPVIYAV FTSSGSVFRG SAVCVYSMAD IRMVFNGPFA HKEGPNYQWM PFSGKMPYPR PGTCPGGTFT PSMKSTKDYP DEVINFMRSH PLMYQAVYPL QRRPLVVRTG APYRLTTIAV DQVDAADGRY EVLFLGTDRG TVQKVIVLPK DDQELEELML EEVEVFKDPA PVKTMTISSK RQQLYVASAV GVTHLSLHRC QAYGAACADC CLARDPYCAW DGQACSRYTA SSKRRSRRQD VRHGNPIRQC RGFNSNANKN AVESVQYGVA GSAAFLECQP RSPQATVKWL FQRDPGDRRR EIRAEDRFLR TEQGLLLRAL QLSDRGLYSC TATENNFKHV VTRVQLHVLG RDAVHAALFP PLSMSAPPPP GAGPPTPPYQ ELAQLLAQPE VGLIHQYCQG YWRHVPPSPR EAPGAPRSPE PQDQKKPRNR RHHPPDT | |
Greater than 90% as determined by SDS-PAGE. |
6405 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |