missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SOX9 Protein
A cDNA sequence encoding the SOX9 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13562-2ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
6662 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MNLLDPFMKM TDEQEKGLSG APSPTMSEDS AGSPCPSGSG SDTENTRPQE NTFPKGEPDL KKESEEDKFP VCIREAVSQV LKGYDWTLVP MPVRVNGSSK NKPHVKRPMN AFMVWAQAAR RKLADQYPHL HNAELSKTLG KLWRLLNESE KRPFVEEAER LRVQHKKDHP DYKYQPRRRK SVKNGQAEAE EATEQTHISP NAIFKALQAD SPHSSSGMSE VHSPGEHSGQ SQGPPTPPTT PKTDVQPGKA DLKREGRPLP EGGRQPPIDF RDVDIGELSS DVISNIETFD VNEFDQYLPP NGHPGVPATH GQVTYTGSYG ISSTAATPAS AGHVWMSKQQ APPPPPQQPP QAPPAPQAPP QPQAAPPQQP AAPPQQPQAH TLTTLSSEPG QSQRTHIKTE QLSPSHYSEQ QQHSPQQIAY SPFNLPHYSP SYPPITRSQY DYTDHQNSSS YYSHAAGQGT GLYSTFTYMN PAQRPMYTPI ADTSGVPSIP QTHSPQHWEQ PVYTQLTRP |
SOX9 Protein | |
Research Use Only | |
SOX9 | |
Human | |
GST | |
SOX9 is supplied in 50mM Tris-Acetate, pH 7.5, 1mM EDTA and 20% Glycerol. |