Learn More
enQuireBio™ Recombinant Human TNFR1 / CD120a / TNFRSF1A Protein
Recombinant Protein
Marque:  enQuireBio™ QP5348-500ug
Informations supplémentaires : Poids : 0.01000kg
 Afficher plus de versions de ce produit
Code produit. 15975939
Quantité Prix
1 2166.23€ / 500µg
3 2036.91€ / 500µg
Date d’expédition estimée: 30-11-2021
Spécification
NP_001056.1
TNFR1 / CD120a / TNFRSF1A Protein
500 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Neutralization of human TNF alpha induced L929 cell cytolysis significantly was induced by less than 100 ng/ml of Human TNFR1 / CD120a / TNFRSF1A which corresponds to an activity of at least 1.0 x 10^4 units/mg.
Human
Untagged
10 mM sodium phosphate, pH 7.5
7132
Greater than 95% as determined by SDS-PAGE.
Research Use Only
TNFRSF1A
Recombinant Protein
E. coli
MDSVCPQGKY IHPQNNSICC TKCHKGTYLY NDCPGPGQDT DCRECESGSF TASENHLRHC LSCSKCRKEM GQVEISSCTV DRDTVCGCKN QYRHYWSENL FQCFNCSLCL NGTVHLSCQE KQNTVCTCHA GFFLRENECV SCSNCKKSLE CTKLCLPQIE N