missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TNFRSF4 Protein
A cDNA sequence encoding the TNFRSF4 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13769-10ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
TNFRSF4 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
LHCVGDTYPS NDRCCHECRP GNGMVSRCSR SQNTVCRPCG PGFYNDVVSS KPCKPCTWCN LRSGSERKQL CTATQDTVCR CRAGTQPLDS YKPGVDCAPC PPGHFSPGDN QACKPWTNCT LAGKHTLQPA SNSSDAICED RDPPATQPQE TQGPPARPIT VQPTEAWPRT SQGPSTRPVE VPGGRALEPK SCDKTHTCPP CPAPELLGGP SVFLFPPKPK DTLMISRTPE VTCVVVDVSH EDPEVKFNWY VDGVEVHNAK TKPREEQYNS TYRVVSVLTV LHQDWLNGKE YKCKVSNKAL PAPIEKTISK AKGQPREPQV YTLPPSRDEL TKNQVSLTCL VKGFYPSDIA VEWESNGQPE NNYKTTPPVL DSDGSFFLYS KLTVDKSRWQ QGNVFSCSVM HEALHNHYTQ KSLSLSPGKH HHHHH | |
Greater than 90.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
TNFRSF4 protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |