missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TNNI3 Protein

A cDNA sequence encoding the TNNI3 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP13777-10ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15974669

Quantité Prix
1 114.77€ / 10µg
10 107.33€ / 10µg
Date d’expédition estimée: 26-11-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!



Description

Description

Spécification

Spécification

TNNI3 Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
TNNI3 protein solution in PBS and 25mM arginine.
Protein is >95% pure as determined by 12% PAGE (coomassie staining).
Research Use Only
Recombinant Protein
E. coli
MADGSSDAAR EPRPAPAPIR RRSSNYRAYA TEPHAKKKSK ISASRKLQLK TLLLQIAKQE LEREAEERRG EKGRALSTRC QPLELAGLGF AELQDLCRQL HARVDKVDEE RYDIEAKVTK NITEIADLTQ KIFDLRGKFK RPTLRRVRIS ADAMMQALLG ARAKESLDLR AHLKQVKKED TEKENREVGD WRKNIDALSG MEGRKKKFES
Vidéos
FDS
Documents
Certificats