Learn More
enQuireBio™ Recombinant Human TSLP Protein
Recombinant Protein
Marque: enQuireBio™ QP5289-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_149024.1 | |
TSLP Protein | |
Research Use Only | |
TSLP | |
Human | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |
85480 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MYDFTNCDFE KIKAAYLSTI SKDLITYMSG TKSTEFNNTV SCSNRPHCLT EIQSLTFNPT AGCASLAKEM FAMKTKAALA IWCPGYSETQ INATQAMKKR RKRKVTTNKC LEQVSQLQGL WRRFNRPLLK QQ | |
Greater than 95% as determined by SDS-PAGE. |