missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TSR2 Protein

A cDNA sequence encoding the TSR2 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP13830-100ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15964839

  • 1740.00€ / 100µg
Expédition estimée: 10-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

90121
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMAGAAED ARALFRAGVC AALEAWPALQ IAVENGFGGV HSQEKAKWLG GAVEDYFMRN ADLELDEVED FLGELLTNEF DTVVEDGSLP QVSQQLQTMF HHFQRGDGAA LREMASCITQ RKCKVTATAL KTARETDEDE DDVDSVEEME VTATNDGAAT DGVCPQPEPS DPDAQTIKEE DIVEDGWTIV RRK
Greater than 90% as determined by SDS-PAGE.
TSR2 Protein
Research Use Only
TSR2
Human
His
TSR2 protein solution (0.5 mg/ml) containing PBS buffer (pH 7.4) and 10% glycerol.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions