missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TYW5 Protein

A cDNA sequence encoding the TYW5 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP13847-1mg

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15974889

  • 3401.86€ / 1mg

Date d’expédition estimée: 30-11-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

TYW5 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
TYW5 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMAGQHLP VPRLEGVSRE QFMQHLYPQR KPLVLEGIDL GPCTSKWTVD YLSQVGGKKE VKIHVAAVAQ MDFISKNFVY RTLPFDQLVQ RAAEEKHKEF FVSEDEKYYL RSLGEDPRKD VADIRKQFPL LKGDIKFPEF FKEEQFFSSV FRISSPGLQL WTHYDVMDNL LIQVTGKKRV VLFSPRDAQY LYLKGTKSEV LNIDNPDLAK YPLFSKARRY ECSLEAGDVL FIPALWFHNV ISEEFGVGVN IFWKHLPSEC YDKTDTYGNK DPTAASRAAQ ILDRALKTLA ELPEEYRDFY ARRMVLHIQD KAYSKNSE
Vidéos
FDS
Documents
Certificats