Learn More
enQuireBio™ Recombinant Human UBE2J2 Protein
A cDNA sequence encoding the UBE2J2 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13862-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
UBE2J2 Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The UBE2J2 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 200mM NaCl, 1mM DTT and 30% glycerol. |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMSSTSS KRAPTTATQR LKQDYLRIKK DPVPYICAEP LPSNILEWHY VVRGPEMTPY EGGYYHGKLI FPREFPFKPP SIYMITPNGR FKCNTRLCLS ITDFHPDTWN PAWSVSTILT GLLSFMVEKG PTLGSIETSD FTKRQLAVQS LAFNLKDKVF CELFPEVVEE IKQKQKAQDE LSSRPQTLPL PDVVPDGETH LVQNGIQLLN GHAPGAVPNL AGLQQANRHH |