missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Ubiquitin K48R Protein

A cDNA sequence encoding the Ubiquitin K48R was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP13871-5ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15974969

Quantité Prix
1 114.77€ / 5µg
10 107.33€ / 5µg
Date d’expédition estimée: 30-11-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!



Description

Description

Spécification

Spécification

7311; 7314; 6233; 7316
Greater than 95% as determined by SDS-PAGE.
Research Use Only
RPS27A; UBB; UBC; UBA52
Human
Untagged
Ubiquitin K48R Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
Diluted in PBS plus 5% glycerol.
Vidéos
FDS
Documents
Certificats