Learn More
enQuireBio™ Recombinant Human UFM1 Protein
A cDNA sequence encoding the UFM1 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13882-5ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
UFM1 Protein | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The UFM1 solution (1 mg/ml) 20mM Tris buffer (pH 8.0) and 10% glycerol. |
Greater than 95.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSKVSFKITL TSDPRLPYKV LSVPESTPFT AVLKFAAEEF KVPAATSAII TNDGIGINPA QTAGNVFLKH GSELRIIPRD RVG |