missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human YY1 Protein
A cDNA sequence encoding the YY1 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13988-20ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
YY1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMASGDTL YIATDGSEMP AEIVELHEIE VETIPVETIE TTVVGEEEEE DDDDEDGGGG DHGGGGGHGH AGHHHHHHHH HHHPPMIALQ PLVTDDPTQV HHHQEVILVQ TREEVVGGDD SDGLRAEDGF EDQILIPVPA PAGGDDDYIE QTLVTVAAAG KSGGGGSSSS GGGRVKKGGG KKSGKKSYLS GGAGAAGGGG ADPGNKKWEQ KQVQIKTLEG EFSVTMWSSD EKKDIDHETV VEEQIIGENS PPDYSEYMTG KKLPPGGIPG IDLSDPKQLA EFARMKPRKI KEDDAPRTIA CPHKGCTKMF RDNSAMRKHL HTHGPRVHVC AECGKAFVES SKLKRHQLVH TGEKPFQCTF EGCGKRFSLD FNLRTHVRIH TGDRPYVCPF DGCNKKFAQS TNLKSHILTH AKAKNNQ | |
Greater than 80.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The YY1 solution (0.25 mg/1ml) contains PBS, 20% glycerol and 1mM DTT. |