missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Zinc finger protein GLI2 Protein
A DNA sequence encoding the Homo sapiens (Human) Zinc finger protein GLI2, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP6088-ye-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_005261.2 | |
Zinc finger protein GLI2 Protein | |
Research Use Only | |
GLI2 | |
Recombinant Protein | |
Yeast | |
EQLADLKEDL DRDDCKQEAE VVIYETNCHW EDCTKEYDTQ EQLVHHINNE HIHGEKKEFV CRWQACTREQ KPFKAQYMLV VHMRRHTGEK PHKCTFEGCS KAYSRLENLK THLRSHTGEK PYVCEHEGCN KAFSNASDRA KHQNRTHSNE KPYICKIPGC TKRYTDPSSL RKHVKTVHGP DAHVTKKQRN DVHLRTPLLK ENGDSEAGTE PGGPESTEAS STSQAVEDCL | |
Greater than 90% as determined by SDS-PAGE. |
2736 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |