Learn More
enQuireBio™ Recombinant Influenza H3N2 Hong Kong Recombinant Protein
A cDNA sequence encoding the H3N2 Hong Kong Recombinant was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12082-2ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
Influenza H3N2 Hong Kong Protein | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Influenza | |
Untagged |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
Insect | |
The Recombinant H3N2 A/ Hong Kong 4801/2014 solution contains 10mM Sodium phosphate, pH 7.4,150mM NaCl and 0.005% Tween-20. |