Learn More
enQuireBio™ Recombinant Influenza H3N2 Wisconsin Protein
A cDNA sequence encoding the H3N2 Wisconsin was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12091-50ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
Influenza H3N2 Wisconsin Protein | |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Influenza | |
Untagged |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
Native Protein | |
The Recombinant H3N2 A/Wisconsin/67/05 solution contains 10mM Sodium phosphate, pH 7.2 and 150mM Sodium Chloride. |