missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Influenza New York 3571/2009 Protein

A cDNA sequence encoding the New York 3571/2009 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP12833-100ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15971689

  • 2610.00€ / 100µg
Expédition estimée: 10-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

Influenza New York 3571/2009 Protein
Research Use Only
Recombinant Protein
Baculovirus
ADLMDTLCIG YHANNSTDTV DTVLEKNVTV THSVNLLEDK HNGKLCKLRG VAPLHLGKCN IAGWILGNPE CESLSTASSW SYIVETSSSD NGTCYPGDFI DYEELREQLS SVSSFERFEI FPKTSSWPNH DSNKGVTAAC PHAGAKSFYK NLIWLVKKGN SYPKLSKSYI NDKGKEVLVL WGIHHPSTSA DQQSLYQNAD AYVFVGSSRY SKKFKPEIAI RPKVRDQEGR MNYYWTLVEP GDKITFEATG NLVVPRYAFA MERNAGSGII ISDTPVHDCN TTCQTPKGAI NTSLPFQNIH PITIGKCPKY VKSTKLRLAT GLRNVPSIQS RSRHHHHHH
Greater than 90% as determined by SDS-PAGE.
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Influenza
His
The H1N1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions