missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse ATP synthase subunit beta, mitochondrial Protein

A DNA sequence encoding the Mus musculus (Mouse) ATP synthase subunit beta, mitochondrial, was expressed in the hosts and tags indicated.

Marque:  enQuireBio™ QP5694-ye-500ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15947069

Quantité Prix
1 1705.00€ / 500µg
3 1590.94€ / 500µg
Expédition estimée
25-11-2020

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!



Spécification

Spécification

NP_058054.2
ATP synthase subunit beta, mitochondrial Protein
500 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
11947
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Atp5b
Recombinant Protein
Yeast
AAQASAAPKA GTATGRIVAV IGAVVDVQFD EGLPPILNAL EVQGRDSRLV LEVAQHLGES TVRTIAMDGT EGLVRGQKVL DSGAPIKIPV GPETLGRIMN VIGEPIDERG PIKTKQFAPI HAEAPEFIEM SVEQEILVTG IKVVDLLAPY AKGGKIGLFG GAGVGKTVLI MELINNVAKA HGGYSVFAGV GERTREGNDL YHEMIESGVI NLKDATSKVA LVYGQMNEPP GARARVALTG LTVAEYFRDQ EGQDVLLFID NIFRFTQAGS EVSALLGRIP SAVGYQPTLA TDMGTMQERI TTTKKGSITS VQAIYVPADD LTDPAPATTF AHLDATTVLS RAIAELGIYP AVDPLDSTSR IMDPNIVGNE HYDVARGVQK ILQDYKSLQD IIAILGMDEL SEEDKLTVSR ARKIQRFLSQ PFQVAEVFTG HMGKLVPLKE TIKGFQQILA GEYDHLPEQA FYMVGPIEEA VAKADKLAEE HGS
Vidéos
Documents