Learn More
enQuireBio™ Recombinant Mouse bFGF / FGF2 Protein
Recombinant Protein
Marque:  enQuireBio™ QP5413-1mg
Informations supplémentaires : Poids : 0.01000kg
 Afficher plus de versions de ce produit
Code produit. 15966079
  • 1885.00€ / 1mg
Expédition estimée: 10-05-2024
pour voir le stock disponible 
Spécification
NP_032032.1
bFGF / FGF2 Protein
Research Use Only
FGF2
Recombinant Protein
E. coli
MPALPEDGGA AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
Greater than 95% as determined by SDS-PAGE.
14173
1 mg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Mouse bFGF / FGF2 induces 3T3 cell proliferation (typical ED50 is ? 0.5 ng/ml).
Mouse
Untagged
10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5