missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Calpain-1 catalytic subunit Protein
A DNA sequence encoding the Mus musculus (Mouse) Calpain-1 catalytic subunit, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP5755-ec-50ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_001103974.1 | |
Calpain-1 catalytic subunit Protein | |
Research Use Only | |
Capn1 | |
Recombinant Protein | |
E. coli | |
MTEELITPVY CTGVSAQVQK KRDKELGLGR HENAIKYLGQ DYETLRARCL QSGVLFQDEA FPPVSHSLGF KELGPHSSKT YGIKWKRPTE LMSNPQFIVD GATRTDICQG ALGDCWLLAA IASLTLNETI LHRVVPYGQS FQDGYAGIFH FQLWQFGEWV DVVIDDLLPT KDGKLVFVHS AQGNEFWSAL LEKAYAKVNG SYEALSGGCT SEAFEDFTGG VTEWYDLQKA PSDLYQIILK ALERGSLLGC SINISDIRDL EAITFKNLVR GHAYSVTGAK QVTYQGQRVN LIRMRNPWGE VEWKGPWSDS SYEWNKVDPY EREQLRVKME DGEFWMSFRD FIREFTKLEI CNLTPDALKS RTLRNWNTTF YEGTWRRGST AGGCRNYPAT FWVNPQFKIR LEEVDDADDY DNRESGCSFL LALMQKHRRR ERRFGRDMET IGFAVYQVPR ELAGQPVHLK RDFFLANASR AQSEHFINLR EVSNRIRLPP GEYIVVPSTF EPNKEGDFLL RFFSEKKAGT QELDDQIQAN LPDEKVLSEE EIDDNFKTLF SKLAGDDMEI SVKELQTILN RIISKHKDLR TNGFSLESCR SMVNLMDRDG NGKLGLVEFN ILWNRIRNYL TIFRKFDLDK SGSMSAYEMR MAIEAAGFKL NKKLHELIIT RYSEPDLAVD FDNFVCCLVR LETMFRFFKL LDTDLDGVVT FDLFKWLQLT MFA | |
Greater than 90% as determined by SDS-PAGE. |
12333 | |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |