missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CST3 Protein

A cDNA sequence encoding the CST3 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP11541-1mg

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15937578

  • 7540.00€ / 1mg
Expédition estimée: 30-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

13010
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MRGSHHHHHH GMASATPKQG PRMLGAPEEA DANEEGVRRA LDFAVSEYNK GSNDAYHSRA IQVVRARKQL VAGVNYFLDV EMGRTTCTKS QTNLTDCPFH DQPHLMRKAL CSFQIYSVPW KGTHSLTKFSCKNA
Greater than 95% as determined by SDS-PAGE.
CST3 Protein
Research Use Only
Cst3
Mouse
His
The sterile filtered concentrated (0.5 mg/ml) protein solution was lyophilized with 20mM Tris and 50mM NaCl pH 7.5.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions