missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse EFNB2 Protein

A cDNA sequence encoding the EFNB2 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP11755-5ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15968258

Quantité Prix
1 171.64€ / 5µg
10 161.27€ / 5µg
Date d’expédition estimée: 30-11-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!



Description

Description

Spécification

Spécification

EFNB2 Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Mouse
His
The EFNB2 solution (0.5 mg/1ml) contains phosphate buffered saline (pH 7.4) and 10% glycerol.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
Insect
RSIVLEPIYW NSSNSKFLPG QGLVLYPQIG DKLDIICPKV DSKTVGQYEY YKVYMVDKDQ ADRCTIKKEN TPLLNCARPD QDVKFTIKFQ EFSPNLWGLE FQKNKDYYII STSNGSLEGL DNQEGGVCQT RAMKILMKVG QDASSAGSAR NHGPTRRPEL EAGTNGRSST TSPFVKPNPG SSTDGNSAGH SGNNLLGSEV ALFALEHHHH HH
Vidéos
FDS
Documents
Certificats