missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Follistatin / FST Protein
Recombinant Protein
Marque: enQuireBio™ QP5428-500ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_001288302.1 | |
Follistatin / FST Protein | |
500 μg | |
Less than or equal to 5.00 EUs/ug as determined by the LAL method. | |
Mouse Follistatin / FST induces MCP-11 cell proliferation (typical ED50 ? 1 ug/ml). | |
Mouse | |
Untagged | |
10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5 |
14313 | |
Greater than 90 as determined by SDS-PAGE. | |
Research Use Only | |
FST | |
Recombinant Protein | |
E. coli | |
MGNCWLRQAK NGRCQVLYKT ELSKEECCST GRLSTSWTEE DVNDNTLFKW MIFNGGAPNC IPCKETCENV DCGPGKKCRM NKKNKPRCVC APDCSNITWK GPVCGLDGKT YRNECALLKA RCKEQPELEV QYQGKCKKTC RDVFCPGSST CVVDQTNNAY CVTCNRICPE PSSSEQYLCG NDGVTYSSAC HLRKATCLLG RSIGLAYEGK CIKAKSCEDI QCGGGKKCLW DSKVGRGRCS LCDELCPDSK SDEPVCASDN ATYASECAMK EAACSSGVLL EVKHSGSCN |