Learn More
enQuireBio™ Recombinant Mouse GDNF Protein
Recombinant Protein
Marque:  enQuireBio™ QP5446-10ug
Informations supplémentaires : Poids : 0.01000kg
Code produit. 15976139
Quantité Prix
1 217.14€ / 10µg
6 204.11€ / 10µg
Date d’expédition estimée: 24-11-2021
Spécification
NP_001288261.1
GDNF Protein
10 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Bioactivity of recombinant GDNF is confirmed by C6 cell proliferation assay (see figure).
Mouse
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA
14573
Greater than 95% as determined by SDS-PAGE.
Research Use Only
GDNF
Recombinant Protein
E. coli
MSPDKQAALP RRENRNRQAA AASPENSRGK GRRGQRGKNR GCVLTAIHLN VTDLGLGYET KEELIFRYCS GSCESAETMY DKILKNLSRS RRLTSDKVGQ ACCRPVAFDD DLSFLDDNLV YHILRKHSAK RCGCI