Learn More
enQuireBio™ Recombinant Mouse GM-CSF / CSF2 Protein
Recombinant Protein
Marque:  enQuireBio™ QP5416-1mg
Informations supplémentaires : Poids : 0.01000kg
Code produit. 15976089
  • 3651.05€ / 1mg
Date d’expédition estimée: 24-11-2021
Spécification
NP_034099.2
GM-CSF / CSF2 Protein
1 mg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Shows bioactivity at a concentration of 50 pg/mL in the FDCP-1 cell proliferation assay. The bioactivity acceptance criteria is set at 2 x 10^7 units/mg.
Mouse
Untagged
10 mM acetic acid
12981
Greater than 95% as determined by SDS-PAGE.
Research Use Only
CSF2
Recombinant Protein
E. coli
MAPTRSPITV TRPWKHVEAI KEALNLLDDM PVTLNEEVEV VSNEFSFKKL TCVQTRLKIF EQGLRGNFTK LKGALNMTAS YYQTYCPPTP ETDCETQVTT YADFIDSLKT FLTDIPFECK KPVQK