Learn More
enQuireBio™ Recombinant Mouse KGF Protein
A cDNA sequence encoding the KGF was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP10734-10ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
KGF Protein | |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Mouse | |
Untagged | |
The protein was lyophilized from a concentrated (1 mg/ml) solution containing 20mM Phosphate buffer pH 8 and 0.1M NaCl. |
Greater than 95.0% as determined by: (a) Analysis by HPLC. (b) Analysis by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MCNDMSPEQT ATSVNCSSPE RHTRSYDYME GGDIRVRRLF CRTQWYLRID KRGKVKGTQE MKNSYNIMEI RTVAVGIVAI KGVESEYYLA MNKEGKLYAK KECNEDCNFK ELILENHYNT YASAKWTHSG GEMFVALNQK GIPVKGKKTK KEQKTAHFLP MAIT |