missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Leptin tA Protein

A cDNA sequence encoding the Leptin tA was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP10762-10ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15975088

Quantité Prix
1 114.77€ / 10µg
10 107.33€ / 10µg
Date d’expédition estimée: 22-11-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!



Description

Description

Spécification

Spécification

Leptin tA Protein
10 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
The protein was lyophilized from a concentrated (0.65 mg/ml) solution with 0.003mM NaHCO3.
Greater than 99.0% as determined by:(a) Gel filtration analysis. (b) Analysis by SDS-PAGE.
Research Use Only
Leptin Antagonist Triple Mutant Mouse Recombinant is capable of inhibiting Leptin-induced proliferation of BAF/3 cells stably transfected with the long form of human Leptin receptor. It also inhibits various Leptin effects in several in vitro bioassays.
Mouse
Untagged
Vidéos
FDS
Documents
Certificats