missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse MCP 1 Protein

A cDNA sequence encoding the MCP 1 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP12663-1mg

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15981149

  • 6230.00€ / 1mg
Expédition estimée: 11-07-2023
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

20296
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Ccl2
Mouse
His
MCP-1 protein solution (0.25 mg/ml) containing PBS (pH 7.4) and 10% glycerol.
MCP 1 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MQPDAVNAPL TCCYSFTSKM IPMSRLESYK RITSSRCPKE AVVFVTKLKR EVCADPKKEW VQTYIKNLDR NQMRSEPTTL FKTASALRSS APLNVKLTRK SEANASTTFS TTTSSTSVGV TSVTVN
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions