missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse MCP-3 / CCL7 Protein
A DNA sequence encoding the Mus musculus (Mouse) MCP-3 / CCL7, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP1492-ec-50ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_038682.1 | |
MCP-3 / CCL7 Protein | |
Research Use Only | |
CCL7 | |
Recombinant Protein | |
E. coli | |
PNASTCCYVK KQKIPKRNLK SYRRITSSRC PWEAVIFKTK KGMEVCAEAH QKWVEEAIAY LDMKTPTPKP | |
Greater than 90% as determined by SDS-PAGE. |
20306 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |