missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse S100A3 Protein

A cDNA sequence encoding the S100A3 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP13393-1mg

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15983419

  • 3915.00€ / 1mg
Expédition estimée: 17-01-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

S100A3 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMTRPLE QAVAAIVCTF QEYAGRCGDK YKICQSELKE LLQKELPTWT PSEFRECDYN KFMSVLDTNK DCEVDFGEYV RSLASLCLYC HEYFKECPPE PPCPQ
Greater than 95.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Mouse
His
S100A3 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer, pH 8.0, 10% glycerol, 2mM DTT and 150mM NaCl.
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions