missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse TGFB3 Protein
A cDNA sequence encoding the TGFB3 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP10876-2ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
21809 | |
2 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The activity is determined by the ability to induce chondrogenic differentiation. | |
Mouse | |
Untagged | |
The Mouse TGFB3 protein solution contains 20% Ethanol and 0.12% Acetic acid. |
TGFB3 Protein | |
Research Use Only | |
Tgfb3 | |
Recombinant Protein | |
E. coli | |
ALDTNYCFRN LEENCCVRPL YIDFRQDLGW KWVHEPKGYY ANFCSGPCPY LRSADTTHST VLGLYNTLNP EASASPCCVP QDLEPLTILY YVGRTPKVEQ LSNMVVKSCK CS | |
Greater than 96.0% as determined by SDS-PAGE. |