missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse TREM2 Protein
A DNA sequence encoding the Mus musculus (Mouse) TREM2, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP4435-10ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_001259007.1 | |
TREM2 Protein | |
Research Use Only | |
Trem2 | |
Recombinant Protein | |
Mammal | |
LNTTVLQGMA GQSLRVSCTY DALKHWGRRK AWCRQLGEEG PCQRVVSTHG VWLLAFLKKR NGSTVIADDT LAGTVTITLK NLQAGDAGLY QCQSLRGREA EVLQKVLVEV LEDPLDDQDA GDLWVPEESS SFEGAQVEHS TSRNQETSFP PTS | |
Greater than 90% as determined by SDS-PAGE. |
83433 | |
10 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |