missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse VEGF (121 a. a. ) Protein
A cDNA sequence encoding the VEGF (121 a. a. ) was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13933-10ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
VEGF (121 a. a. ) Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MAPTTEGEQK SHEVIKFMDV YQRSYCRPIE TLVDIFQEYP DEIEYIFKPS CVPLMRCAGC CNDEALECVP TSESNITMQI MRIKPHQSQH IGEMSFLQHS RCECRPKKDR TKPEKCDKRPR R | |
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
Untagged | |
The protein was lyophilized from a concentrated (1 mg/ml) solution with no additives. |