missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Bet v 1.0101 Protein

A cDNA sequence encoding the Bet v 1.0101 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP11155-1mg

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15976318

  • 5037.65€ / 1mg

Date d’expédition estimée: 23-11-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

Bet v 1.0101 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
His
Bet v 1.0101 protein solution is supplied in 20mM HEPES buffer pH 8.0, 200mM NaCl and 20% glycerol.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
Insect
MGVFNYETET TSVIPAARLF KAFILDGDNL FPKVAPQAIS SVENIEGNGG PGTIKKISFP EGFPFKYVKD RVDEVDHTNF KYNYSVIEGG PIGDTLEKIS NEIKIVATPD GGSILKISNK YHTKGDHEVK AEQVKASKEM GETLLRAVES YLLAHSDAYN
Vidéos
FDS
Documents
Certificats