missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Chimeric Chagas Protein

A cDNA sequence encoding the Chimeric Chagas was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP11408-15ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15997168

  • 381.00€ / 15µg
Expédition estimée: 09-08-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

Chimeric Chagas Protein
15 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
His
Protein is >90% pure as determined by metal affinity chromatography.
Research Use Only
Recombinant Protein
E. coli
Chimeric Chagas Multiantigen was lyophilized from 20mM Tris pH 8.5, 100mM Sodium chloride, 20% trehalose and 0.1% Sodium azide as preservative.
Vidéos
FDS
Documents
Certificats
Promotions

Promotions