missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other GST Protein

A cDNA sequence encoding the GST was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP10656-1mg

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15974538

Quantité Prix
1 2269.63€ / 1mg
3 2133.68€ / 1mg
Date d’expédition estimée: 18-11-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!



Description

Description

Spécification

Spécification

GST Protein
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
MSPILGYWKI KGLVQPTRLL LEYLEEKYEE HLYERDEGDK WRNKKFELGL EFPNLPYYID GDVKLTQSMA IIRYIADKHN MLGGCPKERA EISMLEGAVL DIRYGVSRIA YSKDFETLKV DFLSKLPEML KMFEDRLCHK TYLNGDHVTH PDFMLYDALD VVLYMDPMCL DAFPKLVCFK KRIEAIPQID KYLKSSKYIA WPLQGWQATF GGGDHPPKSD LVPR
Greater than 95% as determined by SDS-PAGE.
Research Use Only
Specific activity is >20 units/mg. A unit is defined as the amount of enzyme that conjugate 1.0 u mole of 1-chloro-2,4-dinitrobenzene (CDNB) with reduced glutathione per minute at pH 6.5 at 25C.
Other
Untagged
GST supplied in Phosphate Buffered Saline pH 7.4.
Vidéos
FDS
Documents
Certificats