missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other LLO Protein

A cDNA sequence encoding the LLO was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP10773-100ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15975118

Quantité Prix
1 1262.51€ / 100µg
3 1186.00€ / 100µg
Date d’expédition estimée: 25-11-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!



Description

Description

Spécification

Spécification

LLO Protein
100 μg
< 1.0 EU per ug protein as determined by the LAL method.
Hemolytic activity for Recombinant Listeriolysin O was 1.18 x 105 HU/mg of protein where HU means hemolytic activity unit that is the amount of toxin needed to release half the hemoglobin (50% lysis) of the erythrocytes as determined by hemolysin assay. 2mM DTT is used to reactivate the toxin.
Other
Untagged
The protein contains 50mM NaH2PO4, 1mM EDTA, 2.7mM KCl, 1mM DTT, 5% (v/v) glycerol and 0.5M NaCl.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
hlyA
Recombinant Protein
E. coli
MKKIMLVFIT LILISLPIAQ QTEAKDASAF HKEDLISSMA PPTSPPASPK TPIEKKHADE IDKYIQGLDY NKNNVLVYHG DAVTNVPPRK GYKDGNEYIV VEKKKKSINQ NNADIQVVNA ISSLTYPGAL VKANSELVEN QPDVLPVKRD SLTLSIDLPG MTNQDNKIVV KNATKSNVNN AVNTLVERWN EKYAQAYPNV SAKIDYDDEM AYSESQLIAK FGTAFKAVNN SLNVNFGAIS EGKMQEEVIS FKQIYYNVNV NEPTRPSRFF GKAVTKEQLQ ALGVNAENPP AYISSVAYGR QVYLKLSTNS HSTKVKAAFD AAVSGKSVSG DVELTNIIKN SSFKAVIYGG SAKDEVQIID GNLGDLRDIL KKGATFNRET PGVPIAYTTN FLKDNELAVI KNNSEYIETT SKAYTDGKIN IDHSGGYVAQ FNISWDEINY DPEGNEIVQH KNWSENNKSK LAHFTSSIYL PGNARNINVY AKECTGLAWE WWRTVIDDRN LPLVKNRNIS IWGTTLYPKY SNSVDNPIE
Vidéos
FDS
Documents
Certificats