missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other PhI p 6 Protein

A cDNA sequence encoding the PhI p 6 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP13043-1mg

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15982329

  • 5800.00€ / 1mg
Expédition estimée: 19-01-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

PhI p 6 Protein
Research Use Only
Recombinant Protein
Baculovirus
MVAMFLAVAV VLGLATSPTA EGGKATTEEQ KLIEDVNASF RAAMATTANV PPADKYKTFE AAFTVSSKRN LADAVSKAPQ LVPKLDEVYN AAYNAADHAA PEDKYEAFVL HFSEALRIIA GTPEVHAVKP GA
Greater than 80.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
His
PhI p 6 is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol.
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions