missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other PMSG Protein

A cDNA sequence encoding the PMSG was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP13081-5000IU

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15962449

  • 161.30€ / 5000 unités

Date d’expédition estimée: 08-11-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

PMSG Protein
Research Use Only
Recombinant Protein
Native Protein
The PMSG was lyophilized with no additives.
5000 IU
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
Vidéos
FDS
Documents
Certificats