Learn More
enQuireBio™ Recombinant other protein 1
A cDNA sequence encoding the Protein A was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13160-50ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
protein 1 | |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MAQHDEAQQN AFYQVLNMPN LNADQRNGFI QSLKDDPSQS ANVLGEAQKL NDSQAPKADA QQNNFNKDQQ SAFYEILNMP NLNEAQRNGFIQSLKDDPSQ STNVLGEAKK LNESQAPKAD NNFNKEQQNA FYEILNMPNL NEEQRNGFIQ SLKDDPSQSA NLLSEAKKLN ESQAPKADNKFNKEQQNAFY EILHLPNLNE EQRNGFIQSL KDDPSQSANL LAEAKKLNDA QAPKADNKFN KEQQNAFYEI LHLPNLTEEQ RNGFIQSLKDDPSVSKEILA EAKKLNDAQA PKEEDNNKPG KEDNNKPGKE DNNKPGKEDG NKPGKEDNKK PGKEDNKKPG KEDNKKPGKE DGNKPGKEDNKKPGKEDGNG VHVVKPGDTV NDIAKANGTT ADKIAADNKL ADKNMIKPGQ ELVVDKKQPA NHADANKAQA LPET |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
spa | |
Other | |
Untagged | |
The Protein-A protein solution contains 20mM Tris-HCl, pH 8 and 10% glycerol. |