missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Shiga Like Toxin 1 Protein

A cDNA sequence encoding the Shiga Like Toxin 1 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP13495-500ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15903759

  • 870.00€ / 500µg
Expédition estimée: 05-06-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

Shiga Like Toxin 1 Protein
Research Use Only
Recombinant Protein
E. coli
Phosphate buffered saline and 50mM arginine, pH 7.4.
500 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
His
Protein is >95% pure as determined by 12% PAGE (coomassie staining).
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions