Learn More
enQuireBio™ Recombinant other TaqDNA Protein
A cDNA sequence encoding the TaqDNA was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13688-1,000U
Informations supplémentaires : Poids : 0.01000kg
Spécification
TaqDNA Protein | |
1000 IU | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MRGMLPLFEP KGRVLLVDGH HLAYRTFHAL KGLTTSRGEP VQAVYGFAKS LLKALKEDGD AVIVVFDAKA PSFRHEAYGG YKAGRAPTPE DFPRQLALIK ELVDLLGLAR LEVPGYEADD VLASLAKKAE KEGYEVRILT ADKDLYQLLS DRIHVLHPEG YLITPAWLWE KYGLRPDQWA DYRALTGDES DNLPGVKGIG EKTARKLLEE WGSLEALLKN LDRLKPAIRE KILAHMDDLK LSWDLAKVRT DLPLEVDFAK RREPDRERLR AFLERLEFGS LLHEFGLLES PKALEEAPWP PPEGAFVGFV LSRKEPMWAD LLALAAARGG RVHRAPEPYK ALRDLKEARG LLAKDLSVLA LREGLGLPPG DDPMLLAYLL DPSNTTPEGV ARRYGGEWTE EAGERAALSE RLFANLWGRL EGEERLLWLY REVERPLSAV LAHMEATGVR LDVAYLRALS LEVAEEIARL EAEVFRLAGH PFNLNSRDQL ERVLFDELGL PAIGKTEKTG KRSTSAAVLE ALREAHPIVE KILQYRELTK LKSTYIDPLP DLIHPRTGRL HTRFNQTATA TGRLSSSDPN LQNIPVRTPL GQRIRRAFIA EEGWLLVALD YSQIELRVLA HLSGDENLIR VFQEGRDIHT ETASWMFGVP REAVDPLMRR AAKTINFGVL YGMSAHRLSQ ELAIPYEEAQ AFIERYFQSF PKVRAWIEKT LEEGRRRGYV ETLFGRRRYV PDLEARVKSV REAAERMAFN MPVQGTAADL MKLAMVKLFP RLEEMGARML LQVHDELVLE APKERAEAVA RLAKEVMEGV YPLAVPLEVE VGIGEDWLSA KE |
Greater than 95.0% as determined by SDS-PAGE. | |
Research Use Only | |
polA | |
Other | |
Untagged | |
Taq DNA Polymerase solution in 20mM Tris-HCl, pH 8.0, 100mM KCl, 0.1mM EDTA, 1mM DTT, 50% Glycerol, 0.5% NP40, 0.5% Tween 20. |