Learn More
enQuireBio™ Recombinant other U. Urealyticum Protein
A cDNA sequence encoding the U. Urealyticum was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13848-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
U. Urealyticum Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged |
Greater than 95% as determined by 10% PAGE (Coomassie staining). | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MB Antigen (2.2 mg/ml) is formulated in 1x PBS and 1mM EDTA pH 7.4 and 0.01% Sodium azide. |