missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Ovine GH Antagonist Protein

A cDNA sequence encoding the GH Antagonist was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP10640-100ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15984488

  • 203.00€ / 100µg
Expédition estimée: 17-01-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

GH Antagonist Protein
Research Use Only
GH G119R acts as antagonist using an in vitro bioassay in PDF-P1 3B9 cells stably transfected with rabbit GH receptors. It is capable of forming a 1:1 complex with the recombinant ovine growth hormone receptor extracellular domain (ECD) and binds to this ECD with affinity similar the the wild type oGH.
Ovine
Untagged
Greater than 98.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE.
100 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.0045mM NaHCO3.
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions