missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rabbit TNC Protein

A cDNA sequence encoding the TNC was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP13765-10mg

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15954629

  • 1330.00€ / 10mg
Expédition estimée: 09-03-2023
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

100009341
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
TNNC2
Rabbit
Untagged
The Troponin C Rabbit protein solution contains 150mM sodium chloride, 10mM sodium phosphate and 0.05% sodium azide, pH 7.2.
TNC Protein
10 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Rabbit
MTDQQAEARS YLSEEMIAEF KAAFDMFDAD GGGDISVKEL GTVMRMLGQT PTKEELDAII EEVDEDGSGT IDFEEFLVMM VRQMKEDAKG KSEEELAECF RIFDRNADGY IDAEELAEIF RASGEHVTDE EIESLMKDGD KNNDGRIDFD EFLKMMEGVQ
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions