missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rabbit TNC Protein

A cDNA sequence encoding the TNC was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP13765-5mg

Informations supplémentaires : Poids : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15974629

Quantità Prezzo
1 701.05€ / 5mg
5 657.80€ / 5mg
Date d’expédition estimée: 24-11-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!



Descrizione

Descrizione

Specifica

Specifica

100009341
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
TNNC2
Rabbit
Untagged
The Troponin C Rabbit protein solution contains 150mM sodium chloride, 10mM sodium phosphate and 0.05% sodium azide, pH 7.2.
TNC Protein
5 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Rabbit
MTDQQAEARS YLSEEMIAEF KAAFDMFDAD GGGDISVKEL GTVMRMLGQT PTKEELDAII EEVDEDGSGT IDFEEFLVMM VRQMKEDAKG KSEEELAECF RIFDRNADGY IDAEELAEIF RASGEHVTDE EIESLMKDGD KNNDGRIDFD EFLKMMEGVQ
Videos
SDS
Documenti
Certificati