missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Allograft inflammatory factor 1 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Allograft inflammatory factor 1, was expressed in the hosts and tags indicated.

Marque:  enQuireBio™ QP5632-ye-100ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15976579

Quantité Prix
1 639.01€ / 100µg
5 601.25€ / 100µg
Date d’expédition estimée: 30-11-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!



Description

Description

Spécification

Spécification

NP_058892.1
Allograft inflammatory factor 1 Protein
100 μg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
29427
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Aif1
Recombinant Protein
Yeast
SQSKDLQGGK AFGLLKAQQE ERLDGINKHF LDDPKYSSDE DLQSKLEAFK TKYMEFDLNG NGDIDIMSLK RMLEKLGVPK THLELKKLIR EVSSGSEETF SYSDFLRMML GKRSAILRMI LMYEEKNKEH QKPTGPPAKK AISELP
Vidéos
FDS
Documents
Certificats