missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat IL 17A/F Protein

A cDNA sequence encoding the IL 17A/F was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP10696-0.1mg

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15974678

Quantité Prix
1 1137.40€ / 100µg
3 1068.13€ / 100µg
Date d’expédition estimée: 26-11-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!



Description

Description

Spécification

Spécification

IL 17A/F Protein
100 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
IL-17A: MAVLIPQSSV CPNAEANNFL QNVKVNLKVL NSLSSKASSR RPSDYLNRST SPWTLSRNED PDRYPSVIWE AQCRHQRCVN AEGKLDHHMN SVLIQQEILV LKREPEKCPF TFRVEKMLVG VGCTCVSSIV RHAS IL-17F: MARRNPKVGL SALQKAGNCP PLEDNSVRVD IRIFNQNQGI SVPRDFQNRS SSPWDYNITR DPDRFPSEIA EAQCRHSGCI NAQGQEDGSM NSVPIQQEIL VLRREPQGCS NSFRLEKMLI KVGCTCVTPI VHHAA
Greater than 98.0% as determined by HPLC and SDS-PAGE.
Research Use Only
The ED50 as determined by its ability to induce of IL-6 production from NIH-3T3 cells, is 15.3-23 ng/ml.
Rat
Untagged
Lyophilized from a concentrated (1 mg/ml) solution containing no additives.
Vidéos
FDS
Documents
Certificats