missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat IL 4 Protein
A cDNA sequence encoding the IL 4 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP10716-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
287287 | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50 as determined in a cell proliferation assay using rat splenocytes is less than 2ng/ml corresponding to a specific activity of 500,000IU/mg. | |
Rat | |
Untagged | |
Lyophilized from a 0.2microm filtered concentrated solution in PBS pH 7.4 and 5% trehalose. |
IL 4 Protein | |
Research Use Only | |
Il4 | |
Recombinant Protein | |
E. coli | |
HGCNDSPLR EIINTLNQVT EKGTPCTEMF VPDVLTATRN TTENELICRA SRVLRKFYFP RDVPPCLKNK SGVLGELRKL CRGVSGLNSL RSCTVNESTL TTLKDFLESL KSILRGKYLQ SCTSMS | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |